Spécifications
- Description:Plastic Surgery Scissors, Premium OR-Grade
- Material:Stainless Steel
- Curvature:Curved
- Sterility:Non sterile
- Length:11.4 cm (4.5")
- Supplier No.:47-1345
- Blade Type:Sharp/Sharp, Smooth
Spécifications
A propos de cet article
Plastic Surgery Scissors are non-ratcheted, finger ring scissors used for cutting and dissecting delicate tisssue in various plastic surgery procedures.
Censitrac Ready OR Grade Plastic Surgery Scissors are industry standard.