Desmopressin Acetate
# de catalogue H-7675.0001BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:16679-58-6
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00133962
- Protein synonyms:Desmopressin|[Deamino1|DDAVP|DArg8] Vasopressin
- Protein/peptide name:Desmopressin Acetate
- Molecular weight:1069.23
- Sequence:3-Mercaptopropionyl-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH₂
- Molecular formula:C₄₆H₆₄N₁₄O₁₂S₂
- Cat. no.:H-7675.0001BA
- Supplier No.:H-7675.0001
Spécifications
A propos de cet article
Desmopressin (DDAVP), as the first vasopressin analog with a very high and very specific antidiuretic effect, has been widely used for different therapeutic purposes. DDAVP also improves human learning and memory processes. CAS Number (desmopressin acetate): 62288-83-9.
For desmopressin see H-7675.
Customers Who Bought This Also Bought