beta-Amyloid (1-40) Trifluoroacetate
# de catalogue 10796-844
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:131438-79-4
- Size:0.1 mg
- Storage conditions:–20 ± 5 °C
- Protein/peptide name:beta-Amyloid (1-40) Trifluoroacetate
- Molecular weight:4329.86
- Molecular formula:C₁₉₄H₂₉₅N₅₃O₅₈S
- Cat. no.:10796-844
Spécifications
A propos de cet article
Customers Who Bought This Also Bought