Ac-Lys-Gln-Leu-Arg-Afc I-2005.0005 Trifluoroacetate
Fournisseur: Bachem Americas
Certificats
A propos de cet article
Hepsin, a membrane-anchored serine protease, is overexpressed in ovarian and prostate cancer and in renal cell carcinoma and thus can serve as prognostic marker. Ac-KQLR-AFC is the optimal substrate for human hepsin, as KQLR / VVNG corresponds to its cleavage site. Excitation at 395-400 nm, emission at 495-505 nm.
Customers Who Bought This Also Bought