H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH Trifluoroacetate
# de catalogue H-7344.0005BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:261969-05-5
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD22377748
- Protein/peptide name:H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH Trifluoroacetate
- Molecular weight:881.09
- Sequence:H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH
- Molecular formula:C₄₄H₆₈N₁₀O₉
- Cat. no.:H-7344.0005BA
- Supplier No.:H-7344.0005
Spécifications
A propos de cet article
CALP-3, VKFGVGFK, acts as a calmodulin agonist. The octapeptide interacts with the calmodulin EF-hand motif, the Ca²⁺-binding site. CALP-3 activates phosphodiesterase in the absence of calcium and inhibits Ca²⁺-mediated cytotoxicity and apoptosis. CALP-3 is more active than CALP-1 (H-8155), By preventing trypsin activation within pancreatiic acinar cells CALP-3 shows potential therapeutic potential in the treatment of pancreatitis.
Customers Who Bought This Also Bought