Goserelin (Free Base), Bachem
# de catalogue H-7296.0005BA
Fournisseur: Bachem Americas
Numéro CAS:
Spécifications
- CAS No.:65807-02-5
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00867894
- Protein/peptide name:Goserelin (free base)
- Molecular weight:1269.43
- Sequence:Pyr-His-Trp-Ser-Tyr-D-Ser(tBu)-Leu-Arg-Pro-Azagly-NH₂
- Molecular formula:C₅₉H₈₄N₁₈O₁₄
- Cat. no.:H-7296.0005BA
- Supplier No.:H-7296.0005
Spécifications
A propos de cet article
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Customers Who Bought This Also Bought