Lung, Tuberculosis (Human) Slide
# de catalogue 470183-260
Fournisseur: Avantor
Spécifications
- Description:Lung, Tuberculosis (Human)
- Cat. no.:470183-260
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-260
- Orientation:Section
Spécifications
A propos de cet article
Illustrates Unhealthy Lung Tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Demonstrates damage in lung tissue resulting from tuberculosis.
Customers Who Bought This Also Bought