H-G-Azido-Abu-Oh
Fournisseur: Bachem Americas
Certificats
A propos de cet article
The methionine isostere γ-azidohomoalanine is fed to bacteria for obtaining “clickable” proteins by recombinant synthesis. Additionally, the azido moiety can serve as a probe for studying protein conformations by IR-spectroscopy.
Customers Who Bought This Also Bought