To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:PDZ Domain Containing 9
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PDZD9
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PDZK-9.\|PDZ domain containing protein 9|PDZD9_HUMAN.|PDZ domain containing 9|PDZD 9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10493-674
- Supplier No.:BS-9632R
Spécifications
A propos de cet article
PDZD9 (PDZ domain containing 9) is a 264 amino acid protein that contains one PDZ (DHR) domain and participates in protein binding. Conserved in chimpanzee, dog, cow, mouse and rat, PDZD9 exists as two alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 16p12.1. Chromosome 16 encodes over 900 genes, approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. Giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth, and the rare disorder Rubinstein-Taybi syndrome, characterized by mental retardation and predisposition to tumor growth and white blood cell neoplasias, are associated with chromosome 16. Crohn's disease, a gastrointestinal inflammatory condition, and systemic lupus erythematosis are also associated with chromosome 16.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDZD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat