To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Inhibin, Beta A
- Clonality:Polyclonal
- Gene ID:3624
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:INHBA
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FRP|EDF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-392
- Supplier No.:BS-9116R
Spécifications
A propos de cet article
Members of the BAGE gene family encode antigens that are recognized by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements. BAGE, also known as B melanoma antigen 1 or cancer/testis antigen 2.1, is a 43 amino acid member of the BAGE family. Generally, BAGE proteins are not expressed in normal tissues, except in testis, but are expressed highly in melanomas, bladder carcinomas, head and neck squamous cell carcinomas, and lung and breast cancer carcinomas. BAGE proteins are not expressed in renal, colorectal and prostatic carcinomas, leukemias and lymphomas.
Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: INHBA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human