To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2L 6
- Clonality:Polyclonal
- Gene ID:9246
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UBE2L6
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:UbcH8|RIG-B|Retinoic acid-induced gene B protein|Ubiquitin/ISG15-conjugating enzyme E2 L6|Ubiquitin carrier protein L6|Ubiquitin-protein ligase L6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-962
- Supplier No.:BS-8369R
Spécifications
A propos de cet article
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells, either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. Ubiquitin conjugating enzyme 8 (UBC8) is an E2 enzyme involved in the ubiquitin pathway for protein degradation. Like other E2 enzymes, UBC8 forms a thioester bond with ubiquitin in an E1-dependent manner. UBC8 binds to the human homolog of Drosophila ariadne (HHARI) and UBC7-associated protein (H7-AP1) as well as double ring-finger protein (Dorfin). UBC8 is enriched in the central nervous system and interacts with Parkin, a RING-finger-containing protein implicated in the pathogenesis of familial Parkinson’s disease. Parkin shares sequence homology with other UBC8 binding proteins such as HHARI and H7-AP1.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: UBE2L6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat