To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Double Homeobox 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:DUX3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:double homeobox 3|Putative double homeobox protein 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10494-138
- Supplier No.:BS-9664R-FITC
Spécifications
A propos de cet article
DUX3 belongs to the paired homeobox family and contains 2 homeobox DNA-binding domains. It expressed in hepatoma Hep3B cells. DUX genes are present in 3.3-kilobase elements, a tandem repeat family scattered in the genome found on the short arms of all acrocentric chromosomes as well as on several other chromosomes. May be functional despite lack of introns and of a poly(A) addition signal. Two different isoforms are known.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUX3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human