Order Entry
ContactUsLinkComponent
Anti-FAM76B Rabbit Polyclonal Antibody (Cy7®)
Anti-FAM76B Rabbit Polyclonal Antibody (Cy7®)
# de catalogue 10494-080
Fournisseur:  Bioss
Anti-FAM76B Rabbit Polyclonal Antibody (Cy7®)
# de catalogue 10494-080
Fournisseur:  Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Family With Sequence Similarity 76, Member B
  • Clonality:
    Polyclonal
  • Gene ID:
    143684
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    FAM76B
  • Conjugation:
    Cy7®
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10494-080
  • Supplier No.:
    BS-9659R-CY7

Spécifications

A propos de cet article

With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and β thalassemia are caused by HBB gene mutations. Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11. The FAM76B gene product has been provisionally designated FAM76B pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM76B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought