To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4)9, E3 ubiquitin protein ligase
- Clonality:Polyclonal
- Gene ID:92979
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MARCH9
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Membrane associated RING finger protein 9|MARCH IX|membrane associated ring finger C3HC4 9|RING finger protein 179|Membrane associated RING CH protein IX|MARH9_HUMAN.|RNF179|E3 ubiquitin protein ligase MARCH9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10489-122
- Supplier No.:BS-9343R-CY7
Spécifications
A propos de cet article
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). MARCH9 (membrane-associated ring finger (C3HC4) 9), also known as RNF179, is a 346 amino acid multi-pass membrane protein that localizes to the golgi apparatus and contains one RING-CH-type zinc finger. Expressed ubiquitously, MARCH9 exists as a homodimer and functions as an E3 ubiquitin-protein ligase that accepts a ubiquitin residue from an E2 ubiquitin-conjugating enzyme and is thought to promote the degradation of target proteins, such as CD4 and MHC-I. Multiple isoforms of MARCH9 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MARCH9
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat