To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4)8, E3 ubiquitin protein ligase
- Clonality:Polyclonal
- Gene ID:220972
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MARCH8
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:MIR|Cellular modulator of immune recognition|RNF178|MARCH-VIII|MARCH8|E3 ubiquitin-protein ligase MARCH8|c-MIR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10493-610
- Supplier No.:BS-9342R-A555
Spécifications
A propos de cet article
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. MARCH8 (membrane-associated ring finger (C3HC4) 8), also known as MIR or RNF178 (RING finger protein 178), is a 291 amino acid multi-pass membrane protein that localizes to vesicle membranes and contains one RING-CH-type zinc finger. Expressed in a variety of tissues, including immature dendritic cells, MARCH8 functions as an E3 ubiquitin ligase that is thought to regulate immune responses by promoting the ubiquitination and subsequent degradation of target proteins, such as B7-2 and CD71.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MARCH8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat