To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 131
- Clonality:Polyclonal
- Gene ID:9867
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RNF131
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10670-474
- Supplier No.:BS-9290R-A350
Spécifications
A propos de cet article
PJA2, also known as E3 ubiquitin-protein ligase praja-2, RNF131 (ring finger protein 131) or Neurodap1, is a 708 amino acid protein that contains one ring-type zinc finger and exists as two alternatively spliced isoforms. Significantly conserved in chimpanzee, dog, cow, mouse, rat, chicken and zebrafish, PJA2 shares 52% identity with PJA1, which is involved in protein ubiquitination in brain and may play a role in X-linked mental retardation. Encoded by a gene that maps to human chromosome 5q21.3, PJA2 localizes to both endoplasmic reticulum and Golgi apparatus membranes. Participating in E2-dependent, E3 ubiquitin-protein ligase activity, PJA2 binds to a variety of E2s and interacts with ubiquitin-conjugating enzymes, such as UBE2D2, in vitro.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF131
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat