To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 25
- Clonality:Polyclonal
- Gene ID:64320
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RNF25
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:AO7|E3 ubiquitin-protein ligase RNF25|RNF25_HUMAN.|RNF 25|E3 ubiquitin protein ligase RNF25|RING finger protein AO7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10669-774
- Supplier No.:BS-9240R-A350
Spécifications
A propos de cet article
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). RNF25 (RING finger protein 25), also known as AO7, is a 459 amino acid protein that contains one RWD domain and one RING-type zinc finger. Via its RING-type zinc finger (a common domain that contains a conserved cysteine-rich region and is found in a number of viral and eukaryotic proteins), RNF25 is thought to exhibit E2-dependent E3 ubiquitin-protein ligase activity. Specifically, the RING finger of RNF25 interacts with an E2 ligase and, through this interaction, facilitates the ubiquitination of target proteins. RNF25 may also regulate the transcriptional activity of NF-kappa-B, a protein that plays important roles in cell survival, inflammation response, host defense and proliferation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF25
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat