To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Serpin Family B Member 10
- Clonality:Polyclonal
- Gene ID:5273
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:SERPINB10
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Serpin B10|Bomapin|PI10|Protease inhibitor 10|Serpin peptidase inhibitor clade B ovalbumin member 10|SPB10_HUMAN.|Protease inhibitor 10 ovalbumin type bomapin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10669-224
- Supplier No.:BS-9211R-A488
Spécifications
A propos de cet article
SERPINB10 is a serine proteinase inhibitor of the ovalbumin like B clade of serpins. It was first discovered in human bone marrow in a search for serpins similar to SerpinB6. SERPINB10 was identified in lymphocytes, and was elevated in chronic myeloid leukemia and chronic myelomonocytic leukemia cells. The monocytic cell lines THP-1 and AML-193 also showed elevated SERPINB10 expression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SERPINB10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human