To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Serpin Peptidase Inhibitor, Clade A (alpha-1 antiproteinase, antitrypsin), Member 10
- Clonality:Polyclonal
- Gene ID:51156
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SERPINA10
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:member 10 |protein Z-dependent protease inhibitor|antitrypsin)|ZPI|serpin peptidase inhibitor|serine (or cysteine) proteinase inhibitor|PZI|SERPINA10 |clade A (alpha-1 antiproteinase|protein Z-dependent protease inhibitorprecursor|member 10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10669-174
- Supplier No.:BS-9209R-A350
Spécifications
A propos de cet article
ZPI, also known as SERPINA10 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10) or PZI, is a 444 amino acid secreted protein that functions as a Protein Z-dependent protease inhibitor. Expressed by the liver, ZPI is secreted into the plasma where, in the presence of calcium, Protein Z and phospholipids, it inhibits the activated pro-coagulation factors X and XI (Factor X and Factor XI). This inhibition helps properly regulate intravenous blood clotting. ZPI, a member of the serpin protein family, contains five potential N-linked glycosylation sites and a tyrosine at position 387 which, when disrupted, renders ZPI inactive. Defects in the gene encoding ZPI may increase susceptibility to venous thrombosis, the formation of blood clots within a vein.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat