To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Centrosome And Spindle Pole Associated Protein 1
- Clonality:Polyclonal
- Gene ID:79848
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CSPP1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CSPP 1|Centrosome and spindle pole associated protein 1|CSPP|CSPP1_HUMAN.|FLJ38886|FLJ22490
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10669-166
- Supplier No.:BS-9208R-FITC
Spécifications
A propos de cet article
Centrosomes are dynamic organelles involved in many aspects of cell function and growth. Centrosomes act as microtubule organizing centers, and provide a site for concerted regulation of cell cycle progression. Duplication of centrosomes occurs once during each cell cycle and requires proper mitotic spindle formation and chromosome segregation. Defects in centrosome duplication or function are linked to many human diseases, including various forms of cancer. The centrosome and spindle pole-associated protein 1 (CSPP1) interacts with centrosomes and microtubules and may play a role in the regulation of G(1)/S-phase progression and spindle assembly. Two isoforms of CSPP1 exist as a result of alternative splicing events. Isoform 1 expression increases throughout the cell cycle and peaks in G2/M phase, whereas isoform 2 expression is highest in G1 phase and decreases thereafter.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CSPP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: