To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Glycerophosphodiester Phosphodiesterase Domain Containing 3
- Clonality:Polyclonal
- Gene ID:79153
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GDPD3
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GDPD3|GDE7|Glycerophosphodiester phosphodiesterase domain-containing protein 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:105-150/318
- Purification:Purified by Protein A
- Cat. no.:10669-116
- Supplier No.:BS-9206R-A488
Spécifications
A propos de cet article
GDE3, also known as glycerophosphodiester phosphodiesterase 3, glycerophosphodiester phosphodiesterase domain containing 2 (GDPD2) or osteoblast differentiation promoting factor (OBDPF), is a 539 amino acid protein belonging to the glycerophosphoryl diester phosphodiesterase family. Possessing glycerophosphoinositol inositolphosphodiesterase activity, GDE3 hydrolyzes glycerophosphoinositol to form inositol 1-phosphate and glycerol, and is suggested to play a role in Actin cytoskeleton remodeling and osteoblast differentiation and growth. A multi-pass membrane protein, GDE3 localizes to cell membrane and cytoplasm, and colocalizes with actin in the cytoskeleton. GDE3 contains one GDPD domain, binds calcium as a cofactor and is encoded by a gene mapping to human chromosome Xq13.1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GDPD3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat