To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Immunoglobulin Superfamily Member 1
- Clonality:Polyclonal
- Gene ID:3547
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:IGSF1
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CHTE|p120|IGDC1|INHBP|PGSF2|IGCD1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10668-856
- Supplier No.:BS-9194R-A647
Spécifications
A propos de cet article
IGSF1 is a highly glycolylated immunoglobulin domain-containing protein. IGSF1 has been shown to act as a coreceptor in inhibin signaling, however, it does not appear to be a high-affinity inhibin receptor by itself. May reduce or inhibit activin A signaling and is believed to be necessary in the mediation of specific effects of inhibin B on activin-stimulated transcription. IGSF1 has been found to interact with several members of the ACVR family and possibly some members of the BMPR group. There are 3 known isoforms of IGSF1, with 1 and 2 likely being multi-pass membrane proteins. Isoform 3 is believed to be expressed as a secreted form. Expression is high in pancreas, testis and fetal liver, while heart, prostate and small intestine show only moderate expression. IGSF1 may be found at very low levels in brain, muscle, thymus, ovary, colon, fetal lung and fetal kidney. Isoform 3 has been detected in pituitary gland.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IGSF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat