To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:U-box Domain Containing 5
- Clonality:Polyclonal
- Gene ID:22888
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UBOX5
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:hUIP5|UIP5|RNF37|UBCE7IP5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-946
- Supplier No.:BS-9145R-A350
Spécifications
A propos de cet article
The U-box domain is a modified RING finger motif that has been implicated in the ubiquitin/proteasome system. The ubiquitin-conjugating enzyme 7-interacting protein 5 (UIP5), also designated U-box domain-containing protein 5 or RING finger protein 37, contains 1 RING-type zinc finger and 1 U-box domain. UIP5 has been shown to interact with UBCH7, an enzyme that mediates selective degradation of abnormal proteins. The gene encoding UIP5 maps to chromosome 20, which houses over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UBOX5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat