To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Zinc And Ring Finger 1, E3 ubiquitin protein ligase
- Clonality:Polyclonal
- Gene ID:84937
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ZNRF1
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Zrfp1|zinc ring finger protein 1|ring finger protein 42|zinc and ring finger 1|NIN283|Rnf42|B830022L21Rik|MGC101991|E3 ubiquitin-protein ligase ZNRF1|nerve injury-induced gene 283 protein|nin283|peripheral nerve injury protein nin283
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10667-838
- Supplier No.:BS-9139R-CY7
Spécifications
A propos de cet article
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. ZNRF1 (zinc and ring finger 1), also known as NIN283, is a 227 amino acid protein that contains one RING-type zinc finger and localizes to the lysosome and the endosome, as well as to cytoplasmic vesicles and the peripheral membrane. Expressed primarily in nervous system tissue, but also present in testis and thymus, ZNRF1 functions as an E3 ubiquitin-protein ligase that is thought to play a role in the establishment and maintenance of neuronal plasticity. Multiple isoforms of ZNRF1 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNRF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat