Order Entry
ContactUsLinkComponent
Anti-AKIRIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-AKIRIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
# de catalogue 10666-758
Fournisseur:  Bioss
Anti-AKIRIN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
# de catalogue 10666-758
Fournisseur:  Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Akirin 2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    AKIRIN2
  • Conjugation:
    Alexa Fluor® 488
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Chromosome 6 Open Reading Frame 166|Fourteen-Three-Three Beta Interactant 1|C6orf166|FBI1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10666-758
  • Supplier No.:
    BS-9082R-A488

Spécifications

A propos de cet article

Required for the innate immune response. Downstream effector of the Toll-like receptor (TLR), TNF and IL-1 beta signaling pathways leading to the production of IL-6. Forms a complex with YWHAB that acts to repress transcription of DUSP1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AKIRIN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought