To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:PDZ Domain Containing 7
- Clonality:Polyclonal
- Gene ID:79955
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PDZD7
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PDZD7_HUMAN.|OTTMUSP00000044305|OTTMUSP00000044304|RP11-108L7.9|PDZ domain containing 7|9130207N01|EG435601|PDZK7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10666-040
- Supplier No.:BS-9042R-A488
Spécifications
A propos de cet article
PDZK7, also known as PDZD7, is a 517 amino acid protein that contains two PDZ (DHR) domains. Encoded by a gene that maps to human chromosome 10q24.31, PDZK7 is conserved in dog, mouse and rat, and exists as three alternatively spliced isoforms. PDZK7 is known to interact with Harmonin, MASS1, USH1G and Usherin. Localizing to nucleus, PDZK7 is expressed in retinal pigment epithelium and inner ear. Biallelic inactivation of PDZK7 can cause non-syndromic hearing impairment and chromosomal aberrations, which are linked to non-syndromic sensorineural deafness. PDZK7 mutations are also linked to Usher syndrome, which is characterized by retinitis pigmentosa and sensorineural deafness, and Alzheimer disease. The gene that encodes PDZK7 maps to human chromosome 10q24.31.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDZD7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat