To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Immunoglobulin Superfamily Containing Leucine-rich Repeat 2
- Clonality:Polyclonal
- Gene ID:57611
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ISLR2
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ISLR2_HUMAN.|Immunoglobulin superfamily containing leucine-rich repeat protein 2|Leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10665-748
- Supplier No.:BS-9025R-CY7
Spécifications
A propos de cet article
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. ISLR2 (immunoglobulin superfamily containing leucine-rich repeat 2), also known as LINX (leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein), is a 745 amino acid single-pass membrane protein that contains five LRR repeats, one Ig-like (immunoglobulin-like) domain, a LRRCT domain and one LRRNT domain. ISLR2 exists as a homomultimer and is essential for axon extension during neural development. The gene encoding ISLR2 maps to human chromosome 15q24.1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ISLR2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat