To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Growth Hormone Releasing Hormone Receptor
- Clonality:Polyclonal
- Gene ID:2692
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GHRHR
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GHRH receptor|Growth hormone-releasing hormone receptor|GHRHR|GRF receptor|GRFR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10671-332
- Supplier No.:BS-8867R-FITC
Spécifications
A propos de cet article
GHRH-R is a seven transmembrane domain protein that localizes to the somatotroph of the pituitary. GHRH-R plays an important role in growth and acts as a high-affinity receptor for GHRH. Binding of GHRH leads to the coupling of GHRH-R to G-protein which stimulates increased adenylyl cyclase activity and the accumulation of cAMP leading to the synthesis and release of growth hormone and somatotroph proliferation. In addition, this signalling pathway may have direct action in fetal/placental development, reproduction and immune function. GHRH and GHRH-R may also play a role in the regulation of non-rapid eye movement sleep (NREMS). The expression of GHRH-R is dependent on the presence of the POU domain factor Pit-1. Mutations in the gene encoding this protein can result in isolated growth hormone deficiency (IGHD), also known as Dwarfism of Sindh, and anterior pituitary hypoplasia (APH).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GHRHR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat