To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ankyrin Repeat Domain 6
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ANKRD6
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ANKR6_HUMAN|Ankyrin repeat domain-containing protein 6|ANKRD 6|ankyrin repeat domain 6|Diversin.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10671-266
- Supplier No.:BS-8728R-HRP
Spécifications
A propos de cet article
Diversin, also known as ANKRD6 (ankyrin repeat domain 6), is a 727 amino acid protein that contains eight ankyrin repeats and belongs to the ankyrin repeat domain protein family. Members of the ankyrin repeat domain family facilitate protein-protein interactions and function as adaptors of signaling pathways. Expressed in a developmentally-regulated manner and at highest levels in the brain, Diversin is believed to play a role in brain development. Via its ankyrin repeats, Diversin can directly interact with Dvl (dishevelled), an interaction that is essential for the activation of noncanonical Wnt signaling. In addition, Diversin contains a C-terminal domain that binds Axin/Conductin and a casein kinase-binding domain in its central region that specifically binds casein kinase Ié. Through the action of these additional domains, Diversin may also facilitate canonical Wnt signaling. Due to alternative splicing events, three Diversin isoforms exist.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ANKRD6
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat