To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Leucine-rich PPR motif-Containing protein
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:LRP130
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GP130|LPPRC_HUMAN.|130 kDa leucine-rich protein|LRPPRC|LRP 130|Leucine-rich PPR-mot containing protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10664-702
- Supplier No.:BS-8719R-A647
Spécifications
A propos de cet article
Leucine-rich protein (LRP130) is a cytoplasmic mRNA-binding protein likely to be involved in the processing of mitochondrial DNA transcripts. Defects in the LRPPRC gene that encodes LRP130 result in the French-Canadian type of Leigh syndrome, a severe neurological disorder characterized by lesions in the subcortical region of the brain. LRP130 also interacts with the low-affinity receptor for leukemia inhibitory factor to produce an intracelluar signal cascade.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRP130
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat