To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Viperin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Viperin
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10664-690
- Supplier No.:BS-8718R-FITC
Spécifications
A propos de cet article
RSAD2 is a 361 amino acid protein that is involved in antiviral defense against pathogens such as Hep C, Cytomegalovirus and HIV-1. Localized to the cytosolic side of the endoplasmic reticulum and relocated to the Golgi apparatus upon viral infection, RSAD2 is thought to prevent viral budding by disrupting lipid rafts at the plasma membrane and supporting the Interferon-induced antiviral state of the cell. Additionally, RSAD2 can bind to and inactivate FDPS (an enzyme that is crucial for the synthesis of cholesterol and geranylated and farnesylated proteins), thereby playing a role in lipid synthesis. Overexpression of RSAD2 leads to abnormal lipid accumulation that is associated with atherosclerosis, a chronic inflammatory disease characterized by hardened arteries.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Viperin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat