To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ETS variant 5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ETV5
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ETS translocation variant 5|ETV5_HUMAN.|Etv5|Ets variant gene 5|Ets related protein ERM|ERM|Ets-related protein ERM
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10663-648
- Supplier No.:BS-8592R-A555
Spécifications
A propos de cet article
Rad and Gem related GTP binding protein (REM) is a member of the RGK subfamily of Ras-like GTPases that also includes Rad, REM2 and Gem/Kir. REM is a phosphorylated protein that is highly expressed in cardiac muscle and moderately expressed in lung, kidney and skeletal muscle. REM associates with several 14-3-3 isoforms as well as with calmodulin in a calcium-dependent manner. REM mediates two distinct signal transduction pathways that regulate both cytoskeletal reorganization and voltage-gated calcium channel activity. REM decreases the current that passes through cardiac voltage-gated L-type Ca channels (Ca(V)). Overexpression of REM may result in the development of cytoplasmic processes, reorganization of the Actin cytoskeleton, reduction in focal adhesion size and an elongated or dendritic-like cell morphology.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ETV5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat