To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Nuclear Factor Of Activated T-Cells, Cytoplasmic, Calcineurin-Dependent 4
- Clonality:Polyclonal
- Gene ID:4776
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:NFATC4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:NF-AT3|cytoplasmic 4|NF-ATc4|NFATc4|Nuclear Factor Activated T-Cells|Nuclear factor of activated T-cells|T cell transcription factor NFAT3
- Modification:phospho-Ser168/Ser170
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10662-628
- Supplier No.:BS-8535R-FITC
Spécifications
A propos de cet article
The product of this gene is a member of the nuclearfactors of activated T cells DNA-binding transcription complex.This complex consists of at least two components: a preexistingcytosolic component that translocates to the nucleus upon T cellreceptor (TCR) stimulation and an inducible nuclear component.Other members of this family of nuclear factors of activated Tcells also participate in the formation of this complex. Theproduct of this gene plays a role in the inducible expression ofcytokine genes in T cells, especially in the induction of the IL-2and IL-4. Alternatively spliced transcript variants encodingdifferent isoforms have been noted for this gene. [provided byRefSeq, Oct 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NFATC4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse