To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2G 1
- Clonality:Polyclonal
- Gene ID:7326
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:UBE2G1
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ubiquitin carrier protein G1|UBC7|Ubiquitin-protein ligase G1|Ubiquitin-conjugating enzyme E2 G1|E217K
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-758
- Supplier No.:BS-8371R-A488
Spécifications
A propos de cet article
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. In Saccharomyces cerevisiae, ubiquitin-like proteins include Rub1, Ula1, Uba3, Smt3, Ubc2, Ubc12 and Ubc9. Rub1 shares 53% homology with ubiquitin and requires activation via the E2 proteins, including Ula1, Uba3 and Ubc12 in order to conjugate to substrates directed to different proteolytic systems. Ubc4 catalyzes ubiquitination of IkBa in a phosphorylation and SCFB-TRCP dependent manner. In this particular reaction, E1 first transfers ubiquitin to the E2 component Ubc4, and Ubc4 then associates with E3 ligase, which conjugates the poly-ubiquitin chain on a target protein. In this fashion, the chain tags the IkBa for degradation by a proteasome thus lifting the inhibitory effect of IkBa on NFkB and allowing NFkB to enter the nucleus.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UBE2G1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat