To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2G 2
- Clonality:Polyclonal
- Gene ID:7327
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UBE2G2
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ubiquitin-protein ligase G2|Ubiquitin-conjugating enzyme E2 G2|UBC7|Ubiquitin carrier protein G2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-486
- Supplier No.:BS-8349R-CY3
Spécifications
A propos de cet article
UBE2G2 (Ubiquitin-conjugating enzyme E2 G2), also known as UBC7, is a 165 amino acid protein involved in ubiquitin-mediated protein degradation. Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). UBE2G2 is an E2 ubiquitin-conjugating enzyme that acts to catalyze the covalent attachment of ubiquitins to various proteins. Expressed throughout the body, UBE2G2 shares 100% sequence identity with its mouse counterpart and is thought to be involved in endoplasmic reticulum-associated degradation (ERAD). Two isoforms of UBE2G2 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UBE2G2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat