To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2D2
- Clonality:Polyclonal
- Gene ID:7322
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UBE2D2
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:UBCH4|E2(17)KB2|UBCH5B|PUBC1|UBC4/5|UBC4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10482-464
- Supplier No.:BS-8348R-CY5
Spécifications
A propos de cet article
Ubiquitin is an abundant, highly conserved protein found in all eukaryotic cells either free or covalently attached to cellular proteins. The primary function of ubiquitin in mammalian systems is to clear abnormal, foreign, and improperly folded proteins by targeting them for proteosome degradation. UBE2D proteins, including UBE2D1 (ubiquitin-conjugating enzyme E2D1 or UBC5A), UBE2D2 (ubiquitin-conjugating enzyme E2D2 or UBC5B) and UBE2D3 (ubiquitin-conjugating enzyme E2D3 or UBC5C), are E2 ubiquitin-conjugating enzymes that catalyze the ubiquitination of I˚Bå in a phosphorylation and SCFB-TRCP-dependent manner. Specifically, E1 first transfers a ubiquitin residue to the E2 component (a UBE2D protein), and the UBE2D protein then associates with an E3 ubiquitin-protein ligase, which immediately transfers that residue to a protein that is targeted for degradation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UBE2D2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat