To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Angiopoietin-like 3
- Clonality:Polyclonal
- Gene ID:27329
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ANGPTL3
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FHBL2|ANG-5|ANL3|ANGPT5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/460
- Purification:Purified by Protein A
- Cat. no.:10426-566
- Supplier No.:BS-5918R-A350
Spécifications
A propos de cet article
Angiopoietin-like protein 3 (Angptl3) functions as a potent lipoprotein lipase inhibitor and is an important component of plasma triglyceride homeostasis. Angptl3 also plays a role in adipose formation and angiogenesis through its interaction with integrin ?v)beta(3). It is secreted by the liver and is functionally defined by the C-terminal fibrinogen (FBN)-like domain and an N-terminal coiled-coil domain. Angptl3 regulates circulating triglyceride levels during different nutritional states thereby mediating the feeding/fasting cycle. A deficiency of Angptl3 results in abnormally low lipid levels, and a repression of the protein may be protective against atherosclerosis. Angptl3 may also play an important role in hyperlipidemia in diabetes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat