To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Adenosine A2b Receptor
- Clonality:Polyclonal
- Gene ID:136
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ADORA2B
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HGNC:264|ADORA2|netrin-1R|adenosine A2bR|ADORA2B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:145-178/332
- Purification:Purified by Protein A
- Cat. no.:10426-140
- Supplier No.:BS-5900R
Spécifications
A propos de cet article
Receptor for adenosine. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase.
Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100
Type: Primary
Antigen: ADORA2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat