To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:H-prolyl hydroxylase 4
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:HPH4
- Conjugation:Cy3®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PH 4|EGLN4|Proline 4 hydroxylase|Prolyl hydroxlase domain containing 4|PH4|P4htm|Transmembrane prolyl 4-hydroxylase.|Prolyl 4 hydroxylase transmembrane endoplasmic reticulum|HPH4|Hypoxia-inducible factor prolyl hydroxylase 4|PHD4|P4H with transmembrane domain|H-prolyl hydroxylase 4|P4HTM_HUMAN|H-PH4|HPH-4|Hypoxia inducible factor prolyl 4 hydroxylase|P4H TM|P4H-TM|H prolyl hydroxylase PH4|Hypoxia inducible factor prolyl hydroxylase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10426-128
- Supplier No.:BS-5899R-CY3
Spécifications
A propos de cet article
Catalyzes the post-translational formation of 4-hydroxyproline in hypoxia-inducible factor (HIF) alpha proteins. Hydroxylates HIF1A at 'Pro-402' and 'Pro-564'. May function as a cellular oxygen sensor and, under normoxic conditions, may target HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitination complex.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HPH4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat