To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Hepatitis C Virus NS5A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Antigen symbol:NS5A
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HCV|NICE-3|HepC NS5|HCV NS5|HepC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10399-130
- Supplier No.:BS-4856R-A488
Spécifications
A propos de cet article
HCV is classified into the genus Hepacivirus of the family Flaviviridae. Like all the members of the family, HCV is an enveloped, single-stranded, positive-sense RNA virus. Its genome (about 9600 nt) is flanked at both termini by conserved, highly structured non-translated regions (NTRs) and encodes a polyprotein precursor (about 3000 aa), which is proteolytically processed by host and viral proteases to produce the structural (core, E1, E2 and p7) and non-structural (NS2, NS3, NS4A, NS4B, NS5A and NS5B) proteins of the virus. Recently, an additional protein has been identified, whose function remains unknown. NS5A is a ~56 kDa pleiotropic protein with key roles in both viral RNA replication and modulation of the physiology of the host cell. It's exact role is not currently known (2008).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NS5A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus