To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Epstein–Barr virus latent membrane protein 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Antigen symbol:LMP2
- Conjugation:Cy7®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HHV4|LMP2_EBVB9 |EBV LMP2B.|EBV LMP2A|Latent membrane protein 2B|Terminal protein|EBV latent membrane protein 2A|Latent membrane protein 2|LMP2|Epstein Barr virus|Human Herpesvirus 4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:400-450/497
- Purification:Purified by Protein A
- Cat. no.:10405-604
- Supplier No.:BS-4700R-CY7
Spécifications
A propos de cet article
Epstein Barr virus (EBV) is a member of the herpesvirus family and one of the most common human viruses. Most people become infected with EBV during their lives. Primary infections usually results in infectious mononucleosis (glandular fever) but the virus can also lay dormant in B lymphocytes and when reactivated become associated with more serious disease such as Burkitt's lymphoma, nasopharyngeal carcinoma and Hodgkin's disease. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LMP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus