To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tubulin Beta 3
- Clonality:Polyclonal
- Gene ID:10381
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TUBB3
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Tubulin beta-3 chain|CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:400-450/450
- Purification:Purified by Protein A
- Cat. no.:10405-492
- Supplier No.:BS-4512R-A647
Spécifications
A propos de cet article
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain. TUBB3 plays a critical role in proper axon guidance and mantainance.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat