To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin 3
- Clonality:Polyclonal
- Gene ID:3562
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:IL3
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Multipotential colony-stimulating factor|P-cell-stimulating factor|MCGF|Hematopoietic growth factor|Mast-cell growth factor|Interleukin-3|IL3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10402-852
- Supplier No.:BS-4510R-CY5
Spécifications
A propos de cet article
The pleiotropic IL3 (Interleukin 3) is a 15kDa cytokine that is primarily secreted by activated T lymphocytes and stimulates the proliferation and differentiation of hematopoietic cells. IL3 acts in hematopoiesis by controlling the production, differentiation, and function of two related white cell populations of the blood, the granulocytes and the monocytes/macrophages. It induces granulocytes, macrophages, mast cells, stem cells, erythroid cells, eosinophils and megakaryocytes and is expressed by activated T cells, mast cells, and natural killer cells. IL3 not only supports growth of both pluripotent stem cells and the more differentiated committed progenitors, but it also stimulates the functional activity of some fully differentiated cells. IL3 has also been shown to protect mast cells from undergoing apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human