To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Bactericidal/permeability-increasing Protein
- Clonality:Polyclonal
- Gene ID:671
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BPI
- Conjugation:HRP (Horseradish Peroxidase)
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CAP 57|bactericidal/permeability-increasing protein|BPI|Bactericidal permeability-increasing protein|BPI_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10402-712
- Supplier No.:BS-4292R-HRP
Spécifications
A propos de cet article
The antimicrobial protein Bactericidal Permeability Increasing protein (BPI) is a 55 kDa protein found in the primary granules of polymorhonuclear leukocytes (PMN). The cytotoxicity action of BPI is limited to gram negative bacteria, reflecting the high affinity of BPI for bacterial LPS. Binding of BPI to live bacteria via LPS causes anti-infective activites: 1) cytotoxicity via sequential damage to bacterial outer and inner lipid membranes, 2) neutralization of gram-negative bacterial LPS, 3) opsonization of bacteria to enhance phagocytosis by neutrophils.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human