Anti-PIH1D1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
# de catalogue 10402-582
Fournisseur: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:PIH1 Domain Containing 1
- Clonality:Polyclonal
- Gene ID:55011
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PIH1D1
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PIH1 domain containing protein 1|PIH1 domain-containing protein 1|PIHD1_HUMAN.|NOP17|PIH1D1|PIH1 domain containing 1|Nucleolar protein 17 homolog
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10402-582
- Supplier No.:BS-4273R-A488
Spécifications
A propos de cet article
No data available.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PIH1D1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought