To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:VAMP (vesicle-associated membrane Protein)-associated Protein B
- Clonality:Polyclonal
- Gene ID:9217
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:VAPB
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ALS8|VAP-B|VAMP-B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10402-302
- Supplier No.:BS-4227R-A647
Spécifications
A propos de cet article
VAPB contains 1 MSP domain and it may play a role in vesicle trafficking. Defects in VAPB are a cause of proximal adult autosomal dominant spinal muscular atrophy [MIM:182980]; also called late onset spinal muscular atrophy Finkel type. Spinal muscular atrophies are neurodegenerative disorders characterized by degeneration of lower motor neurons, leading to progressive paralysis muscular atrophy. This form is a late adult onset form of the disease (after age 20 years). The patients show a benign course, most of them remaining ambulatory 10 to 40 years after clinical onset.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: VAPB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat