To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Clonality:Polyclonal
- Gene ID:7363
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:UGT2B4
- Conjugation:Alexa Fluor® 488
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10402-274
- Supplier No.:BS-4225R-A488
Spécifications
A propos de cet article
The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of the UGT2B family are involved in the metabolism of bile acids, phenol derivatives, catecholestrogens and steroids. Although it is widely recognized that the liver is the major site of glucuronidation, it is now clear that UGT enzymes are also found in extra-hepatic tissues.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: UGT2B4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human