To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Rabphilin 3A-like (without C2 domains)
- Clonality:Polyclonal
- Gene ID:9501
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RPH3AL
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:NOC2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10405-360
- Supplier No.:BS-4183R-A350
Spécifications
A propos de cet article
Rabphilin-3AL (rabphilin-3A-like), also known as RPH3AL or NOC2, is a cytoplasmic Rab GTPase effector. It contains one FYVE-type zinc finger and one Rab-binding (RBD) domain, but unlike its related protein, rabphilin-3A, rabphilin-3AL does not contain any C2 domains. Rabphilin-3AL is expressed in a variety of tissues, with highest levels found in kidney, skeletal muscle, pancreas, liver, ovary, stomach, heart and thyroid. It is believed to play a role regulating calcium-dependent secretory vesicle exocytosis in endocrine and exocrine cells. Via its RBD domain, rabphilin-3AL is capable of binding Rab 27a and, through this interaction, rabphilin-3AL is recruited to dense-core vesicles. With lower affinity, rabphilin-3AL can also bind Rab 3 and Rab 8 with its RBD domain. Through an interaction with Rab 3, rabphilin-3AL can inhibit G-protein signaling in endocrine pancreas and positively regulate insulin secretion. Rabphilin-3AL knockout mice display accumulation of secretory granules and irregular shape in exocrine cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RPH3AL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat