To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Twist Family BHLH Transcription Factor 2
- Clonality:Polyclonal
- Gene ID:117581
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TWIST2
- Conjugation:Alexa Fluor® 555
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:AMS|SETLSS|DERMO1|bHLHa39|FFDD3|BBRSAY
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10405-328
- Supplier No.:BS-4173R-A555
Spécifications
A propos de cet article
Twist2 (Twist homolog 2) is a basic helix-loop-helix (bHLH) transcription factor which acts as a transcriptional repressor. It binds to the E-box consensus sequence 5'-CANNTG-3' and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Efficient DNA binding requires dimerization with another bHLH protein. Twist2 inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TWIST2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat