To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:HMG-CoA reductase
- Clonality:Polyclonal
- Gene ID:3156
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:HMGCR
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Hydroxymethylglutaryl CoA reductase|HMDH|3 hydroxy 3 methylglutaryl Coenzyme A reductase|3-hydroxy-3-methylglutaryl-coenzyme A reductase|LDLCQ3|Hmgcr|HMG CoAR|HMG-CoA reductase|MGC103269|HMG CoA reductase|3 hydroxymethylglutaryl CoA reductase|3H3M
- Modification:phospho-Ser872
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:840-888/888
- Purification:Purified by Protein A
- Cat. no.:10392-394
- Supplier No.:BS-4063R-FITC
Spécifications
A propos de cet article
Transmembrane glycoprotein that is the rate-limiting enzyme in cholesterol biosynthesis as well as in the biosynthesis of nonsterol isoprenoids that are essential for normal cell function including ubiquinone and geranylgeranyl proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HMGCR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat